Learn More
Trajan Scientific and Medical™ Replacement Needles for Agilent Plunger-in-Needle Syringes
Quality needles to fit a SGE syringe in a range of lengths and tip styles.
66.70€ - 73.05€
Product Code | Brand | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Price | Quantity & Availability | ||||
15258583
|
Trajan Scientific and Medical™
034715 |
1 μL | 23 G |
66.70€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
15208583
|
Trajan Scientific and Medical™
033730 |
0.5 μL | 23 to 26 G |
73.05€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
42 mm | |
Cone |