Learn More
Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths
Designed specifically to generate maximum ultrasonic activity. Grant Instruments™ Replacement Basket for Grant Ultrasonic Baths prevent operators coming into contact with chemical solutions.
Brand: Grant Instruments™ XAB10
Additional Details : Weight : 1.00000kg
Description
- Stainless steel construction
- Works in conjunction with lid to create a drip collection unit
- Prevents user from coming into contact with chemical solutions
- Prevents items from resting on the inside of the tank wall
- For use with Grant Ultrasonic Baths
Specifications
XUB10 Ultrasonic Bath | |
Stainless Steel | |
(WxDxH) 467 x 100 x115 mm |