Learn More
Rephile™ RephiDuo U Pack TIX
Product Code. 15459739 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15459739 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15459739
  • ONLINE EXCLUSIVE PRICE
    400.44€
    324.61€ / Pack of 1
    Save 75.83€
    18% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
This product is designed to remove ionic and organic contaminants below trace level to match the water quality required for your application.
Rephile filter pack. Comparable to QPAK00TIX.
Compatible with following Millipore™ systems:
  • Milli-Q Direct™
Only compatible with systems manufactured before 2013.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit