Description
This product is designed to remove ionic and organic contaminates down to trace levels from DI pure water.
RephiDuo U Pack 2 for MilliQ Century™ systems. Comparable to QGARD00D2.
Compatible with following Millipore™ systems:
- Milli-Q Academic™
- Milli-Q Biocel™
- Milli-Q Element™
- Milli-Q Gradient™
- Milli-Q Synthesis™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Water Purification System |
| Type | Cartidge Kit |