Learn More
Rephile™ RephiDuo L Pack TL
Product Code. 15479729 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15479729 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15479729
  • ONLINE EXCLUSIVE PRICE
    466.47€
    378.47€ / Pack of 1
    Save 88.00€
    18% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
This product is designed to remove ionic and organic contaminants down to trace levels from pure water.
RephiDuo L Pack TL for large Elix™ systems. Comparable to QGARDTL01.
Compatible with following Millipore™ systems:
  • Elix Clinical 35/70™
  • Elix 20/35/70/100™
  • RiOs 30/50/100/150/200™
  • Elix Gulfstream Clinical™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit