Description
This cartridge is specially designed to further purify Deionized (DI) water to produce high resistivity ultrapure water.
Rephile filter pack. Comparable to QGARDT2X1.
Compatible with following Millipore™ systems:
- Milli-Q Advantage™
- Milli-Q Reference™
Only compatible with systems manufactured before 2013.
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Water Purification System |
| Type | Cartidge Kit |