Learn More
Thermo Scientific™ Refrigerator and Freezer Door Options
Customize refrigerator and freezer configurations to better suit the needs of the lab with these refrigerator and freezer door options.
Brand:  Thermo Scientific™ 7118
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11914286
  • 403.00€ / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Alert:
Door options must be specified at time of order.
Description
Description
Customize refrigerator and freezer configurations to better suit the needs of the lab with Thermo Scientific™ Refrigerator and Freezer Door Options which include glass and heated glass doors, as well as either right- or left-hand swing.
For use with Thermo Scientific Forma, Jewett, Puffer Hubbard, and Revco series. Solid doors are standard on most refrigerators unless otherwise specified. Optional dual-pane glass doors must be specified when ordering. Contact your Sales Representative for information about heated glass doors.
  • Glass doors standard on selected blood bank, chromatography and pharmacy refrigerators
  • Sliding glass doors standard on all 45.8 cu. ft. (1297L) models
  • Heated glass doors increase amperage and may require next size breaker
Ordering Information:
Door options must be specified at time of order
Specifications
Specifications
Refrigerator and Freezer Door, Glass
1297 L
Glass
4.7 cu. ft. (133L) Forma™, Jewett, Puffer Hubbard™
45.8 cu. ft.
Glass
Right