Learn More
Abeba™ Reflexor™ 6910 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 6910/37
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15692418
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20347:2012 O1 SRC
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3568)
  • Antistatic
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRC
Microfiber
37
Polyurethane
Black
Leather
Slip-resistant
6910