Learn More
Abeba™ Reflexor™ 6910 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand: Abeba™ 6910/41
Additional Details : Weight : 0.85000kg
Description
- CE EN ISO 20347:2012 O1 SRC
- Smooth leather
- Black
- Slip resistance PU outsole sole
- 2 replaceable Reflexor™ insoles (article no. 3565/ 3568)
- Antistatic
- Lining microfiber, respiration absorbing
- Instep area with lacing
- Hand-sewn seam
Specifications
CE EN ISO 20347:2012 O1 SRC | |
Microfiber | |
41 | |
Polyurethane |
Black | |
Leather | |
Slip-resistant | |
6910 |