Learn More
Buerkle™ Reduced Nipple with Outer Thread
Manufactured from polypropylene.
Brand: Buerkle™ 8506-1234
5.75 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.00750kg
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
Nipple Reduced Outer | |
Autoclavable | |
1/2 to 3/4 in. |
EU Foodstuffs | |
Polypropylene | |
13.5 mm |