Learn More
Fisherbrand™ Red Micro PTFE Stir Bar
Fisherbrand™ Red Micro PTFE Stir Bar is an inert and leak-free stirrer for a wide variety of applications where identification is important.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 16255841
  • 7.35€ / Pack of 10
Estimated Shipment: 07-06-2024
Description
  • Micro stirrer bars
  • For use where identification is of prime importance
  • PTFE coatings are not as inert as pure PTFE
  • Individually polished to stay cleaner for longer
Specifications
Specifications
3 mm
Red
Containers & Vessels / Stirrer Hot Plates
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Micro
PTFE encapsulated Alnico V magnet core
10/Pk.
Yes
6 mm