Learn More
Haleco™ Recovery Bags
Product Code. 15155174
Change view
Click to view available options
Color:
Black
Clear
Unit Size:
Pack of 200
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15155174 Clear Pack of 200
12516239 Black Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15155174 Supplier Haleco™ Supplier No. SAC130T
  • ONLINE EXCLUSIVE PRICE
    225.17€
    107.32€ / Pack of 200
    Save 117.85€
    52% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
Color Clear
Material LDPE, HDPE
Dimensions (L x W) 82 x 120cm
Height (Metric) 120 cm
Weight (Metric) 13.76 kg
Product Type Waste Bag
Capacity (Metric) 130 L
Length (Metric) 82 cm