Learn More
MP Biomedicals™ Ready-Red™ Mixture DNA and RNA Extraction
Used to remove traces of phenol from the aqueous phase of organic nucleic acid extractions.
Brand: MP Biomedicals™ 11RYRED001
16.62 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.25000kg Transport : UN number : 1888 Chem class : 6.1 Pack group : III
Description
- Ready-Red™ is a coloured mixture of chloroform and isoamyl alcohol.
- The red colour (non toxic dye), facilitates the visualization and the separation of aqueous and organic phases during nucleic acids purification by means of phenolic extractions.
- The red dye of Ready-Red™ is not water-soluble and does not interfere with the quality of the nucleic acids purified.
- The nucleic acids recovered can be used in all standard experiments of molecular biology such as sequencing, PCR, cloning, transformations, etc.
- Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Specifications
Store at Room Temperature (15 to 30 °C) | |
Chloroform:isoamyl alcohol 24:1 with red dye | |
1 x 250 mL | |
Yes |
Yes | |
CHCl3, C5H12O | |
>99% | |
Solution |
Safety and Handling
- Ready-Red
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use CO2, powder, or water spray to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : chloroform, isoamylalcohol