Learn More
Lenz Laborglasintrumente Reaction Vessels, Spherical, with Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 06008588
Additional Details : Weight : 0.01000kg
Description
- Without graduation
- With laboratory flange LF
- With groove
- Made of DURAN™ tubing
- Other sizes on request
Specifications
10 L | |
420 mm | |
280 mm |
Duran Glass | |
Lab Flange 200 LF |