Learn More
Hamilton™ Non-Filtered Racked Pipette Tips
Delivering outstanding accuracy and precision, guaranteed.
Brand: Hamilton™ 68008-12
Additional Details : Weight : 0.20000kg
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Pre-sterilized or non-sterile
- 96 tips to a rack
Specifications
200 μL | |
Non-Filtered | |
Sterile | |
Virgin Polypropylene |