missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-propanol, 98%, Thermo Scientific Chemicals

Catalog Number 11383146
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
35320-23-1
C3H9NO
75.111
MFCD00064413
BKMMTJMQCTUHRP-GSVOUGTGSA-N
d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
80308
(2R)-2-aminopropan-1-ol
CC(CO)N
This item is not returnable. View return policy
35320-23-1
C3H9NO
75.111
MFCD00064413
BKMMTJMQCTUHRP-GSVOUGTGSA-N
d-alaninol, r---2-amino-1-propanol, r-2-aminopropan-1-ol, 2r-2-aminopropan-1-ol, h-d-alaninol, 2r-2-amino-1-propanol, r-2-amino-1-propanol, 2-aminopropanol,-, r-alaninol
80308
(2R)-2-aminopropan-1-ol
CC(CO)N

(R)-(-)-2-Amino-1-propanol is used as a chiral intermediate for the synthesis of pharmaceutical products and agrochemicals. It acts as additives in health care products and animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-2-Amino-1-propanol is used as a chiral intermediate for the synthesis of pharmaceutical products and agrochemicals. It acts as additives in health care products and animal feed.

Solubility
Miscible with water.

Notes
Air sensitive and hygroscopic. Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Density 0.965
Boiling Point 174°C to 176°C
Flash Point 83°C (181°F)
Refractive Index 1.4493
Quantity 5 g
UN Number UN2735
Beilstein 1718866
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Optical Rotation −22° (c=2 in Ethanol)
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-Amino-1-propanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'