Learn More
Pyrex™ Quickfit™ Metal Retaining Clips
Use with soxhlet extractors, flat flange lids and reaction vessels. Pyrex™ Quickfit™ Metal Retaining Clips apply positive pressure to both surfaces with spring wire design.
Brand: Pyrex™ JC100F
35.46 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.06660kg
Description
Use with soxhlet extractors, flat flange lids and reaction vessels. Pyrex™ Quickfit™ Metal Retaining Clips apply positive pressure to both surfaces with spring wire design.
- For securing flat flange lids to wide neck reaction flasks and culture vessels
- Stainless steel construction
Specifications
1/Pk. | |
Stainless Steel | |
Securing flat flange lids to wide neck reaction flasks and culture vessels | |
FG100 |
Joint Clamp | |
Glassware with flat flange joints | |
Spring wire design | |
100 mm |