Learn More
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper
Manufactured from chemically resistant Pyrex™ borosilicate glass with hexagonal glass.
Brand: Pyrex 3212/18WC
207.10 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 1.30000kg
Description
- Complies with ISO 4788
- Calibrated to Class A tolerances
- Individually tested and supplied with certificate showing exact volume contained at 5 points on the scale
- Conformity H certified
- Printed with individual serial number for batch certification
- Manufactured from chemically resistant Pyrex borosilicate glass with hexagonal glass
- Graduations in white enamel for improved visibility with dark colored solutions
- Supplied with polyethylene stopper
Specifications
Graduated Cylinder | |
Glass | |
Class A | |
545 mm | |
45/40 |
Hexagonal | |
ISO 4788 | |
2000 mL | |
Polyethylene Stopper | |
Polyethylene |