Learn More
Pyrex™ 40mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ borosilicate glass. Pyrex™ 40mm Glass Büchner Filter Funnnel includes an integral sintered glass disc also constructed with Pyrex™ glass. High resistance to most corrosive agents, and does not require filter papers.
Brand: Pyrex 3630/08
218.50 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.47000kg
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions - No filter papers required - Highly resistant to most corrosive reagents including ammonia and concentrated sulfuric acid
- Choice of disc diameters and porosities
Specifications
50 mL | |
Pyrex Glass, Borosilicate Glass | |
For convenient filtration of solutions | |
Integrated filter plate sintered Glass disc | |
4 | |
Buchner filter funnel |
8mm | |
170 mm | |
40mm | |
100mm | |
10 to 16μm |