missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™120mm Diameter Glass Büchner Filter Funnel

Manufactured from chemically resistant Pyrex™ glass. Pyrex™ 120mm Diameter Glass Büchner Filter Funnel with integral sintered glass disc is highly resistant to most corrosive reagents and eliminates the need for filter papers.

Brand:  Pyrex 3720/04M

331.55 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.



Additional Details : Weight : 0.72000kg

 View more versions of this product

Product Code. 12693222

  • 349.00€ / Pack of 2
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • Comprehensive range of Büchner filter funnels with integral sintered glass discs
  • Disc and funnel body manufactured from chemically resistant Pyrex™ glass
  • For convenient filtration of solutions
    - No filter papers required
    - Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Choice of disc diameters and porosities
Specifications

Specifications

1000 mL
Pyrex Glass, Borosilicate Glass
For easy filtering solutions
Integrated filter plate sintered Glass disc
2
Buchner filter funnel
19mm
290 mm
120mm
120mm
40 to 100μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers