Learn More
Fisherbrand™ PVC Transparent Tubing
Product Code. 12345859 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric) Inner:
3 mm
6.5 mm
8 mm
Unit Size:
10m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
12345859 3 mm 10m
12365859 6.5 mm 10m
12375859 8 mm 10m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12345859
  • ONLINE EXCLUSIVE PRICE
    4.45€
    2.96€ / 10m
    Save 1.49€
    33% Off
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
Can be steam sterilized at 121°C.
  • Non-toxic
Specifications
Diameter (Metric) Inner 3 mm
Material PVC
Overall Length (Metric) 10 m
Thickness (Metric) 0.75 mm
Min. Operating Temperature (Metric) 121°C
Packaging Quantity 1 Pack
Color Transparent
Max. Operating Temperature (Metric) 121°C