Learn More
Dostmann Electronic™ PVC Precision Probes
Rich, robust accessory for hand-held meter Series P700 that is ideal for use in many laboratory, scientific, and research environments.
Brand: Dostmann Electronic 6000-1019
Additional Details : Weight : 0.13000kg
Description
- Accessories for hand-held meters Series P700
- Immersion probe Pt100, -200 to +450°C, ±0.03°C, 300x3mm
Specifications
Probe, Precision | |
PVC | |
Class 1/3 | |
300 mm |