Learn More
BRAND™ PTFE/ Polypropylene T-bore Stopcock
PTFE construction with polypropylene valve lever. Provides outstanding chemical resistance. Includes integrated hose nozzles.
Brand:  BRAND™ 88670
60.30 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10079930
  • 65.80€ / Each
Estimated Shipment: 28-05-2024
Description
  • PTFE construction
  • Polypropylene valve
  • With integrated hose nozzle
Specifications
Specifications
4 mm
7 mm
Polypropylene
PTFE
Integrated hose nozzle