Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Product Code. 11888952 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
6 mm
7 mm
8 mm
10 mm
13 mm
15 mm
20 mm
30 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
Unit Size:
Each
Pack of 10
Product Code. 11888952
Edge
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
Diameter (Metric) 2 mm
Length (Metric) 7 mm
Type Stir Bar, Micro