Learn More
BRAND™ PTFE Magnetic Stirring Bars Retriever
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars Retriever has a magnetic core that is fully sealed to prevent leeching.
Brand:  BRAND™ 137720
29.95 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 0.04140kg
 View more versions of this product
Product Code. 10759011
  • 33.80€ / Each
Estimated Shipment: 27-05-2024
Description
  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.
  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.
Specifications
Specifications
8 mm
1
PTFE
350 mm