Learn More
DWK Life Sciences PTFE Key with Thread, with three part retaining device
For use with polished DURAN™ disposable taps.
Brand:  DWK Life Sciences 286220801
314.00 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 12868041
  • 329.00€ / Pack of 25
Estimated Shipment: 29-07-2024
Description
Disposable PTFE valve plugs in combination with the polished DURAN™ disposable valve represent a grease-free alternative. The 3-part tap safety device is already included in the scope of delivery.
  • Made of PTFE with thread and 3-part valve locking device
  • For use with polished DURAN™ one-way taps
  • Grease-free working
Specifications
Specifications
4 mm
No
25/Pk.
PTFE
18.8 NS
For use with polished DURAN™ one-way taps