Learn More
Fisherbrand™ PTFE Joint Adapter, Full Length A Type
Fisherbrand™ PTFE Joint Adapters are grease-free, high vacuum seals for standard taper ground glass joints, with substantial shoulder for ease of handling and repeated use. High vacuum performance is attained using transverse sealing rings.
Brand:  Fisherbrand™
Additional Details : Weight : 0.06000kg
 View more versions of this product
Product Code. 16206021
  • 32.45€ / Each
Estimated Shipment: 12-09-2024
Description
  • Suitable for high vacuum through the use of transverse trapezoidal sealing rings on the contacting surfaces
  • Use up to 280°C
  • Eliminates the need for grease
  • Leakage Rate: <1 x 10-4 torr, litre, sec-1
  • Sleeves are inert
Specifications
Specifications
45/50
Standard Taper Ground Glass Joints
1
Fisherbrand™ PTFE Joint Adapter, Full Length A Type
PTFE
24/40