Learn More
Welch™ PTFE Inline Filter
Product Code. 12967914 Shop All Welch Ilmvac Products
Change view
Click to view available options
Material (Membrane):
PTFE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Membrane) unitSize
12967914 PTFE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12967914 Supplier Welch™ Supplier No. 11255504
Only null left
Add to basket
Edge
Add to basket
Economical in-line particulate filter maintains the cleanliness and proper operation of your vacuum pump. Welch™ PTFE Inline Filter protects your vacuum pump from fine particulates and aerosols to 0.2 microns.
  • Protects vacuum pump from particulates
  • 0.2 micron pore size
  • PTFE filter
  • Polypropylene housing
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material (Membrane) PTFE
Pore Size 0.2 μm
For Use With (Application) Particulate filtration, vacuum pump
Housing Material PP
Quantity 1 Each
Type Inline Filter, DN 8