Learn More
Fisherbrand™ PTFE Crucibles
Product Code. p-8007747 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
25 mL
75 mL
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16235941 5 mL Pack of 5
10416212 25 mL Each
10119811 75 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16235941
Only null left
Add to basket
Add to basket
Guaranteed inert and crack-free. Fisherbrand™ PTFE Crucibles can be used up to 280°C.
  • Isostatically molded in pure PTFE
  • Heavy wall construction
Specifications
Material PTFE
Width (Metric) 27mm
Capacity (Metric) 5 mL
Diameter (Metric) Outer 27mm
Height (Metric) 16mm
Product Type Containers