Learn More
Saint Gobain Life Sciences Chemware™ PTFE Boiling Stones
Product Code. 10027750
Change view
Click to view available options
Material:
PTFE
Unit Size:
450g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10027750 PTFE 450g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10027750 Supplier Saint Gobain Life Sciences Supplier No. D1069103
  • ONLINE EXCLUSIVE PRICE
    48.00€
    30.93€ / 450g
    Save 17.07€
    35% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
  • Ideal for continuous extraction methods and boiling organic liquids
  • Chemical resistant
  • Minimize superheating
  • Easily cleaned and non-contaminating
  • Shows no trace of face acidity or alkalinity
  • Will not scratch or abrade glassware and leave no residue
  • Usable up to 288°C
  • Stones minimize bumping during reflux and distillation
TRUSTED_SUSTAINABILITY
Specifications
Type Boiling Stone
Material PTFE
For Use With (Equipment) Distillation Columns