Learn More
Fisherbrand™ PTFE Blade for Plain Shafts
Product Code. 16295851 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Width (Metric):
52 mm
65 mm
75 mm
90 mm
105 mm
125 mm
150 mm
Unit Size:
Pack of 10
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
16295851 75 mm Pack of 10
16215861 125 mm Pack of 10
16285851 65 mm Pack of 10
16205861 105 mm Pack of 10
16225861 150 mm Pack of 5
16275851 90 mm Pack of 10
16255851 52 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 16295851
  • ONLINE EXCLUSIVE PRICE
    27.90€
    24.79€ / Pack of 10
    Save 3.11€
    11% Off
View all promotions
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals. Cowie Technology™ PTFE Blade for Plain Shafts fit all standard shafts for blades.
  • Made of PTFE
  • Totally inert and non-scratch
  • Available in square end and plain end format
Specifications
For Use With (Equipment) PTFE Shaft Stirrer, Plain Shaft for Blades
For Use With (Application) Stirring in Narrow Neck Vessel
Height (Metric) Blade 25 mm
Quantity 1 Each
Temperature (Metric) -200°C to +280°C
Thickness (Metric) 3 mm
Type Blades for Plain Shafts
Width (Metric) 75 mm