Learn More
Abeba™ Protektor Line 1610 Shoes
Leather upper construction includes reflective material.
Brand:  Abeba™ 1610/36
Additional Details : Weight : 1.07000kg
 View more versions of this product
Product Code. 15697407
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather white
  • Steel toe cap
  • Penetration resistant steel midsole
  • Slip resistance PU outsole sole
  • Antistatic
  • Leather lining
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20345:2011 S3 SRC
Leather
Slip-resistant
1610
White
36
Polyurethane
Steel