Learn More
Abeba™ Protektor Line 1610 Shoes
Leather upper construction includes reflective material.
Brand: Abeba™ 1610/48
Additional Details : Weight : 1.07000kg
Description
- CE EN ISO 20345:2011 S3 SRC
- Smooth leather white
- Steel toe cap
- Penetration resistant steel midsole
- Slip resistance PU outsole sole
- Antistatic
- Leather lining
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S3 SRC | |
Leather | |
Slip-resistant | |
1610 |
White | |
48 | |
Polyurethane | |
Steel |