Learn More
Abeba™ Protektor Line 1602 Shoes
Leather upper construction includes reflective material.
Brand: Abeba™ 1602/46
Additional Details : Weight : 1.10000kg
Description
- CE EN ISO 20345:2011 S2 SRC
- Smooth leather
- Black
- Steel toe cap
- Slip resistance PU outsole sole
- Antistatic
- Leather lining
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S2 SRC | |
Leather | |
Slip-resistant | |
1602 |
Black | |
46 | |
Polyurethane | |
Steel |