Learn More
Abeba™ Protektor Line 1600 Shoes
Leather upper construction includes reflective material.
Brand:  Abeba™ 1600/42
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 15697387
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Smooth leather white
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Antistatic
  • Leather lining
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Leather
Slip-resistant
1600
White
42
Polyurethane
Steel