Learn More
WTW™ ProfiLine™ pH 3310 Portable pH Meter
Portable, waterproof and robust meters for the determination of the parameters pH, ORP, dissolved oxygen, conductivity and BSB
765.00€ - 985.00€
Specifications
Accuracy (mV) | ±0.3, ±1 |
---|---|
Accuracy (pH) | ±0.1, ±0.01, ±0.005 |
Accuracy (Temperature) | ±0.1°C |
For Use With (Application) | For professional field measurements requiring documentation and data transfer |
Range (pH) | -2 to 19.999 |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10186573
|
WTW™
2AA312 |
Data logger and USB interface, carry case with pH electrode SenTix™ 41, buffer solution STP 4 and STP 7, stand, beaker, CD-ROM, driver software for USB, batteries, cable |
985.00€
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10564724
|
WTW™
2AA310 |
Data logger and USB interface, short instruction manual, CD-ROM and batteries, cable |
765.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
The pH 3310 is an elegant combination of portable meter and data logger for all those who want to save measuring series automatically and keep processing them on a computer. The meter includes a built-in data memory for 5000 GLP-compliant entries with an interval-controlled logger. All data can conveniently be transmitted to a computer via the USB interface. The CMC function visualizes the optimum measuring range and thus supports correct measurements. The graphic display with clear text menus allows the comfortable and safe operation of the meter.Specifications
±0.3, ±1 | |
±0.1°C | |
-2 to 19.999 | |
-1200 to 1200, -2500 to 2500 | |
1, 2, 3, 4, 5 Calibration WTW Technical Buffer, DIN/NIST buffers plus 20 additional buffer set | |
Manual 500/5000 automatic | |
IP-66/67 | |
300 g | |
4 x 1.5V AA or 4 x 1.2V NiMH rechargeable | |
-5°C to +105°C | |
Portable pH meter | |
80 W mm | |
180 mm |
±0.1, ±0.01, ±0.005 | |
For professional field measurements requiring documentation and data transfer | |
±0.005 | |
±0.3, ±1mV | |
LCD | |
CE | |
80 W x 180 H mm | |
3 Years | |
All values ±1 digit | |
AA, NiMH | |
1000 hr. | |
80 mm | |
55 mm |