Learn More
SCHOTT AG Lighting and Imaging™ Power Supply
This Power Supply provides power for VisiLED System. Includes 100-240 VAC.
Brand: SCHOTT AG Lighting and Imaging™ 400050
Additional Details : Weight : 0.30000kg
Description
- Power Supply input 100-240 VAC
- 50/60 Hz
- Output 21 V/1.00 A, UL and CE (incl. Med-Norm)
- For use with the VisiLED Controllers
Specifications
Power Supply | |
VisiLED Controllers MC750, MC1100 or MC1500 | |
21 to 24 VDC |