missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Persulfate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Melting Point | 100°C |
Color | White |
pH | 4 to 5 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 270.3g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium persulfate
- Danger
- Oxidising solid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304-IF INHALED:.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'