missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide (Beads or Powder/Certified ACS), Fisher Chemical™

Catalog Number p-32862
Click to view available options
Quantity:
3 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium bromide
[K+].[Br-]
This item is not returnable. View return policy
7758-02-3
BrK
119.00
MFCD00011358
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
253877
CHEBI:32030
potassium bromide
[K+].[Br-]
Boiling Point 1435°C
Melting Point 734°C
Color White
pH 5 to 8.8
Physical Form Solid
Quantity 3 kg
Assay Percent Range ≥99 %
Formula Weight 119
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure <0.01mmHg at 20°C
Decomposition Information >800°C
Chemical Name or Material Potassium Bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'