Learn More
Kartell™ Polyvinyl Chloride Stirring Rods
Minimalize scratching with a rod constructed of PVC for durability. Kartell™ Polyvinyl Chloride Stirring Rods are the perfect substitue for a glass stirring rod.
Brand: Kartell™ 0049900
Additional Details : Weight : 0.80000kg
Description
Minimalize scratching with a rod constructed of PVC for durability. Kartell™ Polyvinyl Chloride Stirring Rods are the perfect substitue for a glass stirring rod.
- Rigid
- Lightweight
- Unbreakable
- Ideal replacement for glass rods
Specifications
7.15 mm | |
White | |
PVC |
Stirring Rod | |
10/Pk. | |
350 mm |