Learn More
Fisherbrand™ Polystyrene Diamond-Shaped Standard Weighing Boat
Product Code. 12962850 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
5 mL
30 mL
100 mL
Color:
Black
White
Unit Size:
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Unit Size
12962850 30 mL Black Pack of 500
12972850 100 mL Black Pack of 500
12952850 5 mL Black Pack of 500
12932840 5 mL White Pack of 500
12387552 100 mL White Pack of 500
11573422 30 mL White Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12962850
  • ONLINE EXCLUSIVE PRICE
    18.45€
    16.33€ / Pack of 500
    Save 2.12€
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Designed for accurate pouring with minimal sample loss. Fisherbrand™ Polystyrene Diamond-Shaped Standard Weighing Boat features a smooth surface that flexes easily. Disposable boats have a stable, flat bottom and can be used in a variety of lab environments and applications.
  • Features a diamond shape that easily flexes for accurate pouring
  • Wide, flat bottom helps resist tipping
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
  • Temperature range -10 to 70°C
  • Can be used with microwave moisture analyzers
  • 500 FDA/food-grade compliant boats per pack
Specifications
Bottom Shape Flat
Capacity (Metric) 30 mL
Color Black
Disposable Single-use
Material Polystyrene
Shape Diamond
Sterility Non-sterile
Type Weighing Boat
For Use With (Equipment) Microwave Moisture Analyzer
Dimensions (L x W x H) 78 x 56 x 14 mm
Length (Metric) 78 mm
Width (Metric) 56 mm
Height (Metric) 14 mm