Learn More
Corning™ Polystyrene Microplates
Designed for in vitro diagnostic assays
Brand: Corning™ 3831BC
Additional Details : Weight : 2.98000kg
Description
The Stripwell™ low volume microplates allow test kit manufacturers to reduce overall costs, with a reduction in the amount of reagents and antibody necessary to perform assays.- Designed with one-half the well area of a 'regular' 8 well strip, providing a much greater surface to volume ratio
- Recommended working volume of 50μL to 125μL
- Designed for in vitro diagnostic assays
- Untreated surface is hydrophobic in nature
- Color coded strips
- Bar coding available on request
Specifications
Includes Barcode | |
With Lid | |
Black | |
1536 | |
0.0128 mL |
Strip Plate | |
32 x 48 | |
Polystyrene | |
Non-treated | |
Flat Bottom (F-well) |