Learn More
Fisherbrand™ Super Slim Extra Fine Pipette Tips
Product Code. 11861753
Change view
Click to view available options
Volume (Metric):
200 to 2500 μL
5 to 500 μL
Length (Metric):
95mm
117mm
Unit Size:
Pack of 1000
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) unitSize
11861753 200 to 2500 μL 95mm Pack of 500
11807402 5 to 500 μL 117mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11861753 Supplier Fisherbrand™ Supplier No. 162064
Add to basket
Add to basket
Use Fisherbrand™ Super Slim Extra Fine Pipette Tips in the laboratory since they are the critical link in ensuring that the correct volume of liquid is aspirated and dispensed and that the liquid is not contaminated in the process.
  • Autoclavable
  • Super slim
  • Polypropylene
Specifications
Volume (Metric) 200 to 2500 μL
Sterility Non-sterile
Color Translucent
Autoclavable Autoclavable
Length (Metric) 95mm
Tip Style Finely Tapered
Product Line Super Slim