Learn More
Fisherbrand™ Polypropylene 100 Place Slide Box
Product Code. 11771476 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue
Red
White
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11771476 Blue Each
11791476 White Each
11781476 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11771476
  • ONLINE EXCLUSIVE PRICE
    15.59€
    4.17€ / Pack of 1
    Save 11.42€
    73% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Store and transport slides safely with durable polypropylene cases. Fisherbrand™ Polypropylene 100 Place Slide Box stores 100 standard-size slides in a secure box with a hinged, latch-lock lid.
  • Molded polypropylene box holds 100 slides in secure slots
  • Hinged lid is securely closed by latch lock
  • Includes slide inventory sheet for easy identification
  • Stackable cases include feet for maximum stability
  • Sturdy cases with no liner
Specifications
Type Slide Storage Box
Slide Capacity 100
Color Blue
Device Holds 100 (25 x 75 mm and 1 x 3 in. Microscope Slides)
Dimensions (L x W x H) 210 x 169 x 37 mm
For Use With (Equipment) 25 x 75 mm and 1 x 3 in. microscope slides
Height (Metric) 37 mm
Includes Slide Identification Card
Length (Metric) 210 mm
Material Polypropylene
Quantity 1 Ea.
Shape Rectangular
Width (Metric) 169 mm