Learn More
Witeg™ Polypropylene Screw Caps
Click to view available options
Color:
Red
Yellow
Description
- Autoclavable up to 140°C
- GL 45 Thread
- Closed
Specifications
Closure Material | Polypropylene |
Closure Size | GL 25 |
Closure Type | Screw Cap |
Product Type | Screw Cap |
Color | Red |
Includes | 15mm Hole |
Material | Polypropylene |
No. per Pack | 10Pack |
Quantity | 10Pack |
Screw Color | Red |