Learn More
Fisherbrand™ Polypropylene Natural Pipet Tip
Product Code. 11587452 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
1 to 250 μL
1 to 300 μL
200 to 1000 μL
250 to 1000 μL
500 to 2500 μL
Unit Size:
Pack of 1000
Pack of 250
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11587452 200 to 1000 μL Pack of 1000
11557452 1 to 250 μL Pack of 1000
11597442 1 to 200 μL Pack of 1000
11567452 1 to 300 μL Pack of 1000
11597452 250 to 1000 μL Pack of 1000
11507462 500 to 2500 μL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11587452
Add to basket
Edge
Add to basket
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Natural Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Volume (Metric) 200 to 1000 μL
Color Natural