Learn More
Multiroir™ Polypropylene Drawer
Brand: Multiroir™ 12104
Additional Details : Weight : 0.59400kg
Description
Order Info
dimMaterial
Specifications
Transparent | |
Drawer | |
243 x 155 x 64 mm | |
155 mm |
Polypropylene | |
1 Each | |
243 mm | |
64 mm |