Learn More
Kartell™ Polyoxymethylene Clamps
Manufactured from polyoxymethylene for low friction and excellent dimensional stability. Kartell™ Polyoxymethylene Clamps are color coded according to joint size and provide a safe connection for glass conical joints.
Brand:  Kartell™ 3142300
Additional Details : Weight : 0.02070kg
 View more versions of this product
Product Code. 15885577
  • 32.90€ / Pack of 10
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Plastic construction generates a corrosion proof device that does not scratch glass
  • Available in various colors that correspond with the joint size of the connection
  • Autoclavable
  • Safe connecction for standard-finish glass conical joints
  • Plastic construction generates a corrosion proof device that does not scratch glass
  • Available in various colors that correspond with the joint size of the connection
  • Autoclavable
  • Safe connecction for standard-finish glass conical joints
Specifications
Specifications
1/Pk.
POM
Autoclavable
Joint Clamp
Standard-finish glass conical joint size: 14/23
Yellow