Learn More
Kartell™ Polyoxymethylene Clamps
Manufactured from polyoxymethylene for low friction and excellent dimensional stability. Kartell™ Polyoxymethylene Clamps are color coded according to joint size and provide a safe connection for glass conical joints.
Brand: Kartell™ 3293200
Additional Details : Weight : 0.07740kg
Description
- Plastic construction generates a corrosion proof device that does not scratch glass
- Available in various colors that correspond with the joint size of the connection
- Autoclavable
- Safe connecction for standard-finish glass conical joints
- Plastic construction generates a corrosion proof device that does not scratch glass
- Available in various colors that correspond with the joint size of the connection
- Autoclavable
- Safe connecction for standard-finish glass conical joints
Specifications
1/Pk. | |
POM | |
Autoclavable |
Joint Clamp | |
Standard-finish glass conical joint size 29/32 | |
Red |