Learn More
Vitlab™ Polymethylpentene Forceps
Product Code. 15751996
Change view
Click to view available options
Length (Metric):
115 mm
145 mm
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15751996 145 mm Pack of 10
10350881 115 mm Each
11717202 115 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15751996 Supplier Vitlab™ Supplier No. 67995
  • ONLINE EXCLUSIVE PRICE
    21.45€
    17.15€ / Pack of 10
    Save 4.30€
    20% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Manufactured with PMP for very good resilience. VITLab™ Polymethylpentene Forceps are ideal for use in bottling and decanting.
  • White, Pointed, Elastic 
TRUSTED_SUSTAINABILITY
Specifications
Product Type Forceps
Length (Metric) 145 mm
Material PMP
For Use With (Application) For autoclaving and microwaves, bottling and decanting
Quantity 10/Pack
Tip Style Pointed